Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Haao | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020076.2 | NP_064461.1 | 3 - 4 | 331 | 154 | |
XM_006239684.2 | XP_006239746.1 | 3 - 4 | 1236 | 154 | ||
GenBank mRNA | BC085739.1 | - | 3 - 4 | 285 | 7108551 | 154 |
D44494.1 | - | 3 - 4 | 332 | 154 | ||
FQ219702.1 | - | 3 - 4 | 345 | 154 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |