Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankmy2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108019.1 | NP_001101489.1 | 6 - 7 | 942 | 113 | |
XM_006240042.2 | XP_006240104.1 | 6 - 7 | 3327 | 113 | ||
XM_006240043.2 | XP_006240105.2 | 7 - 8 | 3644 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |