Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rdm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105842.1 | NP_001099312.1 | 6 - 7 | 777 | 152 | |
GenBank mRNA | FQ215272.1 | - | 6 - 7 | 710 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Rattus norvegicus 1700 Probe spans exons |