Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Copz2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108294.1 | NP_001101764.1 | 7 - 8 | 543 | 68 | |
GenBank mRNA | FQ225734.1 | - | 7 - 8 | 547 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |