Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Seli | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134754.1 | NP_001128226.2 | 4 - 5 | 401 | 81 | |
GenBank mRNA | BC168235.1 | - | 4 - 5 | 401 | 7444430 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |