Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgf16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021867.3 | NP_068639.1 | 2 - 3 | 379 | 124 | |
GenBank mRNA | AB002561.1 | - | 2 - 3 | 379 | 124 | |
BC085738.1 | - | 653 | 7189721 | 124 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons |