Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Qtrtd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006248320.2 | XP_006248382.1 | 8 - 9 | 1160 | 109 | |
XM_008768759.1 | XP_008766981.1 | 8 - 9 | 1199 | 109 | ||
XM_008768760.1 | XP_008766982.1 | 7 - 8 | 986 | 109 | ||
XM_008768761.1 | XP_008766983.1 | 7 - 8 | 947 | 109 | ||
XM_008768762.1 | XP_008766984.1 | 6 - 7 | 817 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |