Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alkbh6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127450.1 | NP_001120922.1 | 4 - 5 | 268 | 91 | |
GenBank mRNA | BC082048.1 | - | 4 - 5 | 268 | 91 | |
CB581428.1 | - | 4 - 5 | 274 | 91 |
Gene Symbol: LOC100912458 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006228800.2 | XP_006228862.1 | 4 - 5 | 306 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |