Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exosc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107493.2 | NP_001100963.2 | 5 - 6 | 702 | 93 | |
XM_006228437.2 | XP_006228499.1 | 6 - 7 | 743 | 93 | ||
GenBank mRNA | BC167023.1 | - | 5 - 6 | 614 | 93 | |
FQ225297.1 | - | 5 - 6 | 626 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons |