Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btbd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011932.1 | NP_001011932.1 | 4 - 5 | 924 | 128 | |
GenBank mRNA | BC085850.1 | - | 4 - 5 | 924 | 7189751 | 128 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |