Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006229414.2 | XP_006229476.1 | 11 - 12 | 2507 | 80 | |
XM_006229415.2 | XP_006229477.1 | 11 - 12 | 2507 | 80 | ||
XM_008759585.1 | XP_008757807.1 | 11 - 12 | 2510 | 80 | ||
XM_218828.8 | XP_218828.5 | 11 - 12 | 2510 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Probe spans exons |