Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cln8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007686.1 | NP_001007687.1 | 2 - 3 | 711 | 58 | |
XM_006253394.2 | XP_006253456.1 | 2 - 3 | 985 | 58 | ||
GenBank mRNA | BC078982.1 | - | 2 - 3 | 711 | 7112147 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |