Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defb51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037547.1 | NP_001032636.1 | 1 - 2 | 140 | 114 | |
GenBank mRNA | AY621373.1 | - | 1 - 2 | 140 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |