Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191562.1 | NP_001178491.1 | 2100 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Ion Channel Rattus norvegicus Probe spans exons |