Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108925.2 | NP_001102395.1 | 4 - 5 | 283 | 149 | |
GenBank mRNA | BC158732.1 | - | 4 - 5 | 283 | 9092746 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |