Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013032.3 | NP_037164.3 | 10 - 11 | 1277 | 66 | |
GenBank mRNA | AF038571.1 | - | 3 - 4 | 390 | 66 | |
BC061743.1 | - | 10 - 11 | 1277 | 5599027 | 66 | |
D63772.1 | - | 1308 | 66 | |||
U21104.1 | - | 10 - 11 | 1289 | 66 | ||
U39555.1 | - | 10 - 11 | 1367 | 66 | ||
X94255.1 | - | 10 - 11 | 1334 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Rattus norvegicus Probe spans exons |