Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk2ap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109498.1 | NP_001102968.1 | 3 - 4 | 394 | 158 | |
XM_006230903.2 | XP_006230965.1 | 3 - 4 | 332 | 158 | ||
GenBank mRNA | FQ234216.1 | - | 3 - 4 | 158 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons |