Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tollip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109668.1 | NP_001103138.1 | 4 - 5 | 714 | 110 | |
XM_006230607.2 | XP_006230669.1 | 5 - 6 | 795 | 110 | ||
XM_008760003.1 | XP_008758225.1 | 4 - 5 | 659 | 110 | ||
GenBank mRNA | AB185285.1 | - | 1 - 2 | 167 | 110 | |
BC133067.1 | - | 4 - 5 | 600 | 7193007 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |