Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012542.2 | NP_036674.1 | 1 - 2 | 186 | 105 | |
GenBank mRNA | J02852.1 | - | 1 - 2 | 165 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |