Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ethe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106234.1 | NP_001099704.1 | 3 - 4 | 428 | 116 | |
GenBank mRNA | BC158848.1 | - | 3 - 4 | 388 | 7444454 | 116 |
FQ209618.1 | - | 3 - 4 | 399 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |