Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172029.2 | NP_742026.1 | 6 - 7 | 598 | 65 | |
GenBank mRNA | BC081709.1 | - | 6 - 7 | 598 | 7135576 | 65 |
FQ217438.1 | - | 6 - 7 | 636 | 65 | ||
U05821.1 | - | 6 - 7 | 617 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |