Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trafd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053760.2 | NP_446212.2 | 10 - 11 | 1717 | 150 | |
XM_006249376.2 | XP_006249438.1 | 10 - 11 | 1773 | 150 | ||
XM_008769209.1 | XP_008767431.1 | 10 - 11 | 1820 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |