Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107153.1 | NP_001100623.1 | 3 - 4 | 502 | 74 | |
XM_008769337.1 | XP_008767559.1 | 3 - 4 | 510 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |