Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wbscr17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025112.1 | NP_001020283.1 | 7 - 8 | 1271 | 96 | |
XM_008769132.1 | XP_008767354.1 | 7 - 8 | 1139 | 96 | ||
GenBank mRNA | AB040671.1 | - | 7 - 8 | 1271 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |