Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc64 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191667.1 | NP_001178596.1 | 4 - 5 | 910 | 80 | |
XM_008769234.1 | XP_008767456.1 | 4 - 5 | 975 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |