Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhof | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001073389.5 | XP_001073389.2 | 4 - 5 | 1160 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons |