Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105916.1 | NP_001099386.1 | 9 - 10 | 1146 | 95 | |
XM_006249116.2 | XP_006249178.1 | 8 - 9 | 1098 | 95 | ||
XM_006249117.1 | XP_006249179.1 | 9 - 10 | 1114 | 95 | ||
XM_006249118.1 | XP_006249180.1 | 9 - 10 | 1117 | 95 | ||
XM_008769125.1 | XP_008767347.1 | 8 - 9 | 1227 | 95 | ||
XM_008769126.1 | XP_008767348.1 | 8 - 9 | 1057 | 95 | ||
XM_008769127.1 | XP_008767349.1 | 8 - 9 | 1098 | 95 | ||
XM_008769128.1 | XP_008767350.1 | 8 - 9 | 1098 | 95 | ||
GenBank mRNA | BC166451.1 | - | 8 - 9 | 1211 | 8371649 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |