Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcolce | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019237.1 | NP_062110.1 | 7 - 8 | 1143 | 73 | |
GenBank mRNA | AB008534.1 | - | 7 - 8 | 1130 | 73 | |
AF016503.1 | - | 7 - 8 | 1137 | 73 | ||
BC086572.1 | - | 8 - 9 | 1199 | 7304047 | 73 | |
FQ209381.1 | - | 7 - 8 | 1181 | 73 | ||
U94710.1 | - | 7 - 8 | 1143 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |