Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adprhl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013054.1 | NP_001013072.1 | 1 - 2 | 243 | 84 | |
XM_006253439.2 | XP_006253501.1 | 1 - 2 | 717 | 84 | ||
GenBank mRNA | BC083773.1 | - | 1 - 2 | 243 | 7193406 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |