Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bag4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025130.1 | NP_001020301.1 | 2 - 3 | 948 | 120 | |
GenBank mRNA | BC083608.1 | - | 2 - 3 | 948 | 7133487 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Rattus norvegicus Probe spans exons |