Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012027.1 | NP_001012027.1 | 1 - 2 | 95 | 112 | |
GenBank mRNA | BC088467.1 | - | 1 - 2 | 95 | 7127992 | 112 |
FQ209388.1 | - | 1 - 2 | 113 | 112 | ||
FQ209598.1 | - | 1 - 2 | 81 | 112 | ||
FQ210275.1 | - | 1 - 2 | 78 | 112 | ||
FQ210716.1 | - | 1 - 2 | 114 | 112 | ||
FQ210762.1 | - | 1 - 2 | 113 | 112 | ||
FQ211013.1 | - | 1 - 2 | 119 | 112 | ||
FQ211135.1 | - | 1 - 2 | 113 | 112 | ||
FQ218285.1 | - | 1 - 2 | 107 | 112 | ||
FQ218922.1 | - | 1 - 2 | 124 | 112 | ||
FQ218964.1 | - | 1 - 2 | 78 | 112 | ||
FQ219233.1 | - | 1 - 2 | 78 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |