Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fmo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144737.2 | NP_653338.2 | 6 - 7 | 916 | 97 | |
XM_008769604.1 | XP_008767826.1 | 5 - 6 | 1180 | 97 | ||
XM_008769605.1 | XP_008767827.1 | 6 - 7 | 990 | 97 | ||
GenBank mRNA | AF458414.1 | - | 6 - 7 | 883 | 97 | |
BC070904.1 | - | 6 - 7 | 916 | 7100688 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |