Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019336.1 | NP_062209.1 | 2 - 3 | 277 | 67 | |
XM_006249974.2 | XP_006250036.1 | 2 - 3 | 461 | 67 | ||
GenBank mRNA | BC098681.1 | - | 2 - 3 | 277 | 7385088 | 67 |
FQ234088.1 | - | 2 - 3 | 175 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |