Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3h11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047902.1 | NP_001041367.1 | 8 - 9 | 1011 | 147 | |
XM_008769482.1 | XP_008767704.1 | 6 - 7 | 788 | 147 | ||
GenBank mRNA | DQ480752.1 | - | 8 - 9 | 1011 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |