Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfwd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025126.1 | NP_001020297.1 | 5 - 6 | 712 | 117 | |
XM_006250113.2 | XP_006250175.1 | 12 - 13 | 1911 | 117 | ||
XM_006250114.2 | XP_006250176.1 | 12 - 13 | 1899 | 117 | ||
XM_006250115.2 | XP_006250177.1 | 11 - 12 | 1851 | 117 | ||
XM_006250116.2 | XP_006250178.1 | 11 - 12 | 1839 | 117 | ||
GenBank mRNA | BC091284.1 | - | 5 - 6 | 712 | 7304516 | 117 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |