Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmco1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009631.1 | NP_001009631.1 | 258 | 85 | ||
GenBank mRNA | AF531432.1 | - | 181 | 85 | ||
BC088319.1 | - | 258 | 7325542 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |