Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130409.2 | NP_569093.2 | 3 - 4 | 404 | 106 | |
XM_008769531.1 | XP_008767753.1 | 3 - 4 | 601 | 106 | ||
XM_008769532.1 | XP_008767754.1 | 3 - 4 | 600 | 106 | ||
XM_008769533.1 | XP_008767755.1 | 3 - 4 | 601 | 106 | ||
GenBank mRNA | AJ320522.1 | - | 378 | 106 | ||
BC083174.1 | - | 413 | 7096787 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |