Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufs3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106489.1 | NP_001099959.1 | 6 - 7 | 804 | 80 | |
GenBank mRNA | FQ212516.1 | - | 6 - 7 | 628 | 80 | |
FQ219879.1 | - | 6 - 7 | 631 | 80 | ||
FQ229748.1 | - | 6 - 7 | 651 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Rattus norvegicus 1700 Probe spans exons |