Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfyve19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034948.1 | NP_001030120.1 | 9 - 10 | 1069 | 83 | |
XM_006234787.2 | XP_006234849.1 | 9 - 10 | 1008 | 83 | ||
GenBank mRNA | BC099795.1 | - | 9 - 10 | 1069 | 7936116 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |