Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bmp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017178.1 | NP_058874.1 | 1 - 2 | 60 | 88 | |
GenBank mRNA | L20678.1 | - | 1 - 2 | 60 | 88 | |
Z25868.1 | - | 1 - 2 | 60 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Inventoried Probe spans exons |