Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Idh3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053581.1 | NP_446033.1 | 6 - 7 | 549 | 86 | |
XM_006235048.2 | XP_006235110.1 | 6 - 7 | 586 | 86 | ||
GenBank mRNA | BC079113.1 | - | 6 - 7 | 549 | 7126029 | 86 |
FQ214820.1 | - | 6 - 7 | 547 | 86 | ||
FQ215180.1 | - | 6 - 7 | 553 | 86 | ||
FQ215184.1 | - | 6 - 7 | 541 | 86 | ||
FQ215350.1 | - | 6 - 7 | 549 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |