Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ypel4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024369.1 | NP_001019540.1 | 1 - 2 | 180 | 106 | |
GenBank mRNA | BC083749.1 | - | 1 - 2 | 180 | 7192422 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |