Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttbk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107766.1 | NP_001101236.1 | 9 - 10 | 1070 | 115 | |
XM_006234833.2 | XP_006234895.1 | 11 - 12 | 1503 | 115 | ||
XM_006234834.2 | XP_006234896.1 | 11 - 12 | 1146 | 115 | ||
XM_006234835.2 | XP_006234897.1 | 10 - 11 | 1429 | 115 | ||
XM_006234836.2 | XP_006234898.1 | 9 - 10 | 1281 | 115 | ||
XM_006234837.2 | XP_006234899.1 | 11 - 12 | 1149 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Protein Kinase Rattus norvegicus Probe spans exons |