Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nup62cl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006234254.2 | XP_006234316.1 | 3 - 4 | 466 | 93 | |
XM_006234255.2 | XP_006234317.1 | 3 - 4 | 360 | 93 | ||
XM_006234257.2 | XP_006234319.1 | 3 - 4 | 495 | 93 | ||
XM_008773468.1 | XP_008771690.1 | 3 - 4 | 414 | 93 | ||
XM_008773470.1 | XP_008771692.1 | 3 - 4 | 331 | 93 | ||
GenBank mRNA | BC160918.1 | - | 3 - 4 | 440 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Rattus norvegicus Probe spans exons |