Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddrgk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106512.1 | NP_001099982.1 | 4 - 5 | 937 | 57 | |
GenBank mRNA | FQ214225.1 | - | 4 - 5 | 522 | 57 | |
FQ231424.1 | - | 4 - 5 | 561 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |