Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ripply1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008773499.1 | XP_008771721.1 | 1 - 2 | 234 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |