Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271346.1 | NP_001258275.1 | 2 - 3 | 432 | 83 | |
XM_006234498.2 | XP_006234560.1 | 2 - 3 | 425 | 83 | ||
XM_006234499.2 | XP_006234561.1 | 2 - 3 | 465 | 83 | ||
GenBank mRNA | FQ228020.1 | - | 2 - 3 | 432 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |