Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc36a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108127.1 | NP_001101597.1 | 3 - 4 | 285 | 90 | |
XM_006242558.2 | XP_006242620.1 | 3 - 4 | 908 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Transporter Rattus norvegicus Probe spans exons |