Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014087.1 | NP_001014109.2 | 1 - 2 | 81 | 99 | |
XM_008765934.1 | XP_008764156.1 | 1 - 2 | 197 | 99 | ||
GenBank mRNA | BC085333.1 | - | 1 - 2 | 81 | 7114511 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |