Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014087.1 | NP_001014109.2 | 2 - 3 | 162 | 144 | |
XM_008765934.1 | XP_008764156.1 | 2 - 3 | 278 | 144 | ||
GenBank mRNA | BC085333.1 | - | 2 - 3 | 162 | 7114511 | 144 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |